Skip to Content
Merck
All Photos(1)

Key Documents

69838-M

Sigma-Aldrich

DNA Ligation Kit - Novagen

Dependable ligation with maximum flexibility

Synonym(s):

Ligation Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.54

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

dry ice
wet ice

storage temp.

−20°C

General description

The DNA Ligation Kit provides the necessary components for convenient, reproducible ligation of DNA fragments. Separate solutions of all components are provided to allow maximum flexibility and stability when stored at -20°C. The kit is optimized and tested for PCR cloning, ligation of cDNA and PCR products into plasmid and phage lambda vectors, and for linker ligations. One DNA Ligation Kit contains components for 50 blunt-end or single-base overhang ligations, or up to 500 2- to 4-base cohesive-end ligations.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service