Skip to Content
Merck
All Photos(1)

Key Documents

681672

Sigma-Aldrich

Wnt/β-catenin Inhibitor, Cardamonin

The Wnt/β-catenin Inhibitor, Cardamonin, also referenced under CAS 19309-14-9, controls the biological activity of Wnt/β-catenin. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Wnt/β-catenin Inhibitor, Cardamonin, (E)-1-(2,4-dihydroxy-6-methoxyphenyl)-3-phenylprop-2-en-1-one, Wnt Pathway Inhibitor VII, Cardamonin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O4
CAS Number:
Molecular Weight:
270.28
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

orange-yellow to amber

solubility

DMSO: 100 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O(C)c1c(c(cc(c1)O)O)C(=O)\C=C\c2ccccc2

InChI

1S/C16H14O4/c1-20-15-10-12(17)9-14(19)16(15)13(18)8-7-11-5-3-2-4-6-11/h2-10,17,19H,1H3/b8-7+

InChI key

NYSZJNUIVUBQMM-BQYQJAHWSA-N

General description

Cardamonin, a known 2′,4′-dihydroxy-6′-methoxychalcone, was recently identified as an inhibitor of melanocyte differentiation, among many other pharmacology activities. Cardamonin promoted degradation of intracellular β-catenin while suppressing melanogenesis by down-regulating the expression of MITF and tyrosinase, resulting in decreased melanin content. Furthermore, it was found that cardamonin promoted the degradation of β-catenin through a GSK-3β-independent mechanism, which suggested that Siah-dependent pathway and/or a PKCa-dependent pathway, rather than a GSK-3&beta:-dependent pathway, may mediate intracellularb-catenin degradation by cardamonin.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service