Skip to Content
Merck
All Photos(1)

Documents

567530

Sigma-Aldrich

Sodium hydroxide

pellets

Synonym(s):

Sodium hydroxide, NaOH, ‘Caustic soda’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352316
NACRES:
NA.21

product name

Sodium Hydroxide, Pellets,

vapor density

>1 (vs air)

Quality Level

vapor pressure

<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)

Assay

≥95% (titration)

form

pellets (crystalline)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

technique(s)

DNA extraction: suitable

color

white

mp

318 °C (lit.)

solubility

water: soluble

density

2.13 g/cm3 at 20 °C

cation traces

heavy metals (as Pb): ≤0.0005%

shipped in

ambient

storage temp.

10-30°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Used to neutralize acids, hydrolyze fats, and precipitate alkaloids and metals from aqueous solutions.

Application


  • Protein N-linked Homocysteine Determination: Employing sodium hydroxide pellets in a one-pot sample preparation procedure for the analysis of protein N-linked homocysteine using HPLC-FLD, demonstrating its critical role in chemical synthesis and laboratory applications (Piechocka and Głowacki, 2023).

  • Algae Pyrolysis for Hydrogen Production: The use of sodium hydroxide pellets in a molten state combined with Na2CO3 for efficient hydrogen production from algae pyrolysis. This application underscores its potential in renewable energy sectors and environmental sustainability efforts (Li et al., 2023).

  • Hydrothermal Synthesis of ZnO Superstructures: Research into the temperature and pH optimization using sodium hydroxide pellets for the hydrothermal synthesis of ZnO superstructures. This application is crucial for material science, particularly in developing nanomaterials with specific morphological properties (Ejsmont and Goscianska, 2023).

Warning

Toxicity: Corrosive (L)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service