Skip to Content
Merck
All Photos(1)

Key Documents

565725-M

Sigma-Aldrich

SecinH3

A cell-permeable triazolo compound that acts as a Sec7 domain-binding, selective antagonist against cytohesin family small GEFs.

Synonym(s):

SecinH3, N-(4-(5-(1,3-Benzodioxol-5-yl)-3-methoxy-1H-1,2,4-triazol-1-yl)phenyl)-2-(phenylthio)acetamide, Cytohesin Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H20N4O4S
CAS Number:
Molecular Weight:
460.51
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

S(CC(=O)Nc2ccc(cc2)[n]3nc(nc3c4cc5c(cc4)OCO5)OC)c1ccccc1

InChI

1S/C24H20N4O4S/c1-30-24-26-23(16-7-12-20-21(13-16)32-15-31-20)28(27-24)18-10-8-17(9-11-18)25-22(29)14-33-19-5-3-2-4-6-19/h2-13H,14-15H2,1H3,(H,25,29)

InChI key

QPGYAMIHXLCFTJ-UHFFFAOYSA-N

General description

A cell-permeable triazolo compound that acts as a Sec7 domain-binding (Kd ≤250 nM for Sec7 from human cytohesins 1-3), selective antagonist against cytohesin family small GEFs (IC50 ≤5.6 µM in ARF1 GDP/GTP exchange assays using human cytohesin 3, as well as murine and Drosophila homologues). It inhibits large GEFs only at much higher concentrations (IC50 ≥65 µM in GDP/GTP exchange assays using hEFA6-S7 and yGea2-S7) and exhibits little activity against a panel of 10 commonly studied kinases or hSosII. Shown to effectively inhibit insulin signaling both in HepG2 cells in vitro and in Drosophila and mice in vivo.

Biochem/physiol Actions

Kd ≤250 nM for Sec7 from human cytohesins 1-3; IC50 ≤5.6 µM in ARF1 GDP/GTP exchange assays using human cytohesin 3, as well as murine and Drosophila homologues
Primary Target
Cytohesin

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hafner, M., et al. 2006. Nature444, 941.
Fuss, B., et al. 2006. Nature444, 945.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrogel-based biomaterials for cell delivery and tissue regeneration applications are discussed.

Hydrogel-based biomaterials for cell delivery and tissue regeneration applications are discussed.

Hydrogel-based biomaterials for cell delivery and tissue regeneration applications are discussed.

Hydrogel-based biomaterials for cell delivery and tissue regeneration applications are discussed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service