Skip to Content
Merck
All Photos(1)

Key Documents

559304

Sigma-Aldrich

Anti-SAPK/JNK Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
polyclonal
application:
species reactivity:
mouse, rat, human
citations:
4

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

mouse, rat, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPK8(5599)

General description

Protein A and immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~54 kDa SAPK/JNK protein.
Recognizes the ~54 kDa SAPK/JNK protein in uv-treated HEK293 cells.
This Anti-SAPK/JNK Rabbit pAb is validated for use in Immunoblotting, Immunocytochemistry for the detection of SAPK/JNK.

Immunogen

Human
a full-length, recombinant, human p54 SAPK/JNK2 fusion protein

Application

Immunoblotting (1:1000)

Immunocytochemistry (1:200)

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 10 mM HEPES, 50% glycerol, 0.01% BSA, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
UV treated HEK293 cells

Other Notes

Gupta, S., et al. 1996. EMBO J.15(11), 2760.
Coso, O.A., et al. 1995. Cell81, 1137.
Derijard, B., et al. 1994. Cell76, 1025.
Kyriakis, J.M., et al. 1994. Nature369, 156.
Hibi, M., et al. 1993. Genes Dev.7, 2135.
Kyriakis, J.M. and Avruch, J. 1990. J. Biol. Chem.265, 17355.
Recognizes SAPK/JNK regardless of the phosphorylation state. Variables associated with assay conditions will dictate the proper working dilution.



Recommended Protocol for Immunoblotting



Solutions and Reagents

•Transfer Buffer: 25 mM Tris base, 0.2 M glycine, 20% methanol, pH 8.5.

•SDS Sample Buffer: 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 10% glycerol, 50 mM DTT, 0.1% bromphenol blue.

•10X TBS (Tris-buffered saline): To prepare 1 liter, 24.2 g Tris base, 80 g NaCl, adjust pH to 7.6 with HCl. Dilute 1:10 for use.

•Blocking Buffer: 1X TBS, 0.1% Tween®-20 detergent with 5% non-fat dry milk.

•Primary Antibody Dilution Buffer: 1X TBS, 0.1% Tween-20 detergent with 5% BSA

•Wash Buffer (TBST): 1X TBS, 0.1% Tween-20 detergent



Blotting Membrane

Nitrocellulose or PVDF membranes may be used.



Protein Blotting

A general protocol for sample preparation using 2x106 293 cells per well in a 6-well plate is as follows:



1. Aspirate media. Treat cells by adding fresh media containing regulator for desired time.

2. Aspirate media from cultures; wash cells with PBS; aspirate.

3. Lyse cells by adding 100 µl of SDS Sample Buffer and immediately scrape the cells off the plate and transfer the extract to a microfuge tube. Keep on ice.

4. Sonicate for 2 s to shear DNA and reduce sample viscosity.

5. Heat sample to 95-100°C for 5 min. Cool on ice.

6. Microcentrifuge for 5 min.

7. Load 20 µl onto SDS-PAGE gel (10 cm x 10 cm).

8. Electrotransfer to nitrocellulose membrane.



As controls, we recommend using 20 µl lysate from UV treated HEK293 cell.



Membrane Blocking, Gel and Antibody Incubations

1. After transfer, wash membrane with 25 ml TBS for 5 min at room temperature.

2. Incubate membrane in 25 ml Blocking Buffer for 1-3 h at room temperature or overnight at 4°C.

3. Wash 3 times for 5 min each with 15 ml TBST.

4. Incubate membrane and primary antibody (at the appropriate dilution) in 10 ml Primary Antibody Dilution Buffer with gentle agitation overnight at 4°C.

5. Wash 3 times for 5 min each with 15 ml TBST.

6. Incubate membrane with conjugated secondary antibody at the appropriate dilution in 10 ml Blocking Buffer with gentle agitation for 1 h at room temperature.

7. Wash membrane as in step 5.



Detection of Proteins

Chemiluminescence.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Verstraelen et al.
Cellular and molecular gastroenterology and hepatology, 18(1), 89-104 (2024-04-01)
Mounting evidence suggests the gastrointestinal microbiome is a determinant of peripheral immunity and central neurodegeneration, but the local disease mechanisms remain unknown. Given its potential relevance for early diagnosis and therapeutic intervention, we set out to map the pathogenic changes
Vincenzo Giansanti et al.
Journal of cellular and molecular medicine, 17(1), 103-115 (2012-12-05)
The pathogenesis of age-related macular degeneration (AMD) involves demise of the retinal pigment epithelium and death of photoreceptors. In this article, we investigated the response of human adult retinal pigmented epithelial (ARPE-19) cells to 5-(N,N-hexamethylene)amiloride (HMA), an inhibitor of Na(+)
Kyubin Lee et al.
Biology, 9(8) (2020-08-13)
Abeliophyllum distichum Nakai is known as a monotypic genus endemic to South Korea. Currently, several pharmacological studies have revealed that A. distichum extract exhibits diverse biological functions, including anti-cancer, anti-diabetic, anti-hypertensive, and anti-inflammatory activities. In this study, we present the
Aveline Filliol et al.
Scientific reports, 7(1), 9205-9205 (2017-08-25)
Hepatocyte death is a central event during liver disease progression, in which immune cells play key roles by activating members of the Tumor Necrosis Factor Receptor Superfamily (TNFRSF), including TNFR1 (TNFRSF1A), Fas (TNFRSF6) and TRAIL-R2 (TNFRSF10B). Receptor Interacting Protein Kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service