Skip to Content
Merck
All Photos(1)

Key Documents

553511

Sigma-Aldrich

Rac1 Inhibitor II

The Rac1 Inhibitor II, also referenced under CAS 1090893-12-1, controls the biological activity of Rac1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Rac1 Inhibitor II, 3-((3,5-Dimethylisoxazol-4-yl)methoxy)-N-(4-methyl-3-sulfamoyl-phenyl)benzamide, Z62954982

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21N3O5S
CAS Number:
Molecular Weight:
415.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥93% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable isoxazolyl-benzamide compound that is more effective than NSC23766 (Cat. Nos. 553502 & 553508) in inhibiting PDGF-BB- (Cat. No. 521225) induced cellular Rac1 activation (45.8% vs 11.1% inhibition by 4 h pretreatment of the respective compound at 50 µM) in serum-starved SMCs (human aortic smooth muscle cells) by interfering Rac1-Tiam1 interaction, while exhibiting no effect toward cellular Cdc42 and RhoA activation or Rac1 interaction to its effector Pak1. Shown to effectively prevent PDGF-BB-induced membrane ruffling and lamellipodia formation in serum-starved 3T3 cells (4 h pretreatment at 25 µM). Also available in InSolution format (Cat. No. 553512).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keita Hanada et al.
Cancer letters, 522, 129-141 (2021-09-21)
Mutations of KRAS gene are found in various types of cancer, including colorectal cancer (CRC). Despite intense efforts, no pharmacological approaches are expected to be effective against KRAS-mutant cancers. Macropinocytosis is an evolutionarily conserved actin-dependent endocytic process that internalizes extracellular
Yueyang Wang et al.
eLife, 12 (2023-09-19)
Cell spreading and migration play central roles in many physiological and pathophysiological processes. We have previously shown that MFN2 regulates the migration of human neutrophil-like cells via suppressing Rac activation. Here, we show that in mouse embryonic fibroblasts, MFN2 suppresses
Wenqing Zhou et al.
Journal of cell science, 133(17) (2020-08-14)
Neutrophils rely on glycolysis for energy production. How mitochondria regulate neutrophil function is not fully understood. Here, we report that mitochondrial outer membrane protein Mitofusin 2 (MFN2) regulates neutrophil homeostasis and chemotaxis in vivo Mfn2-deficient neutrophils are released from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service