Skip to Content
Merck
All Photos(1)

Key Documents

5.33965

Sigma-Aldrich

IAP Antagonist, BV6

Synonym(s):

IAP Antagonist, BV6, (S,S,2S,2ʹS)-N,Nʹ-((2S,2ʹS)-(hexane-1,6-diylbis(azanediyl))bis(3-oxo-1,1-diphenylpropane-3,2-diyl))bis(1-((S)-2-cyclohexyl-2-((S)-2-(methylamino)propanamido)acetyl)pyrrolidine-2-carboxamide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C70H96N10O8
CAS Number:
Molecular Weight:
1205.57
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

SMILES string

N2([C@@H](CCC2)C(=O)N[C@@H](C(c8ccccc8)c7ccccc7)C(=O)NCCCCCCNC(=O)[C@@H](NC(=O)[C@H]5N(CCC5)C(=O)[C@@H](NC(=O)[C@@H](NC)C)C6CCCCC6)C(c4ccccc4)c3ccccc3)C(=O)[C@@H](NC(=O)[C@@H](NC)C)C1CCCCC1

InChI key

DPXJXGNXKOVBJV-YLOPQIBLSA-N

General description

A bivalent SMAC mimetic that antagonizes cIAP1 and XIAP interaction with initiator caspases and triggers their proteasomal degradation. Shown to rapidly deplete levels of cIAP1 and XIAP in HCC193 (at 1 µM) and H460 (at 5 µM) lung cancer cell lines. Levels of cIAP1 are reduced within an hour following treatment while XIAP depletion takes up to 24 h. Effectively reduces the viability of HCC193 (IC50 = 7.2 µM) and H460 (IC50 = 30 µM) cells and synergistically enhances their sensitivity to radiation treatment. Reported to increase apoptosis via caspase-8 activation in HCC193 cells (~ 1 µM) and via caspase-9 in H460 cells (~ 5 µM). Also shown to increase the secretion of TNFa in HCC193 cells in a dose-dependent manner. Also enhances cytokine-induced killer (CIK) cell-mediated cytotoxicity against multiple hematological (H9, THP-1, Tanoue) and solid malignant cells (RH1, RH30, and TE671).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
IAP
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Rettinger, E., et al. 2014. Front in Pediatr.2, 75.
Li, W., et al. 2011. J. Thorac. Oncol.6, 1801.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service