523350
PTIO
Nitric oxide radical scavenger.
Synonym(s):
PTIO, 2-Phenyl-4,4,5,5-tetramethylimidazoline-1-oxyl-3-oxide
About This Item
Recommended Products
Quality Level
Assay
≥98% (titration)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
dark blue
solubility
ethanol: soluble
water: soluble
shipped in
ambient
storage temp.
−20°C
SMILES string
[N+]1(=C([NH](=O)C(C1(C)C)(C)C)c2ccccc2)[O-]
InChI
1S/C13H17N2O2/c1-12(2)13(3,4)15(17)11(14(12)16)10-8-6-5-7-9-10/h5-9H,1-4H3
InChI key
DYUUGILMVYJEHY-UHFFFAOYSA-N
General description
Biochem/physiol Actions
Nitric oxide radical scavenger
Warning
Reconstitution
Other Notes
Maeda, H., et al. 1994. Jpn. J. Cancer Res.85, 331.
Yoshida, M., et al. 1994. Biochem. Biophys. Res. Commun.202, 923.
Akaike, T., et al. 1993. Biochemistry32, 827.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service