Skip to Content
Merck
All Photos(1)

Key Documents

5.08506

Sigma-Aldrich

Gabapentin

Synonym(s):

Gabapentin, L-type Voltage Gated Ca2+ Channel Blocke, Gabapentin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17NO2
CAS Number:
Molecular Weight:
171.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 100 mM

storage temp.

2-8°C

SMILES string

[N+H3]CC1(CCCCC1)CC(=O)[O-]

InChI

1S/C9H17NO2/c10-7-9(6-8(11)12)4-2-1-3-5-9/h1-7,10H2,(H,11,12)

InChI key

UGJMXCAKCUNAIE-UHFFFAOYSA-N

General description

A water soluble GABA analog originally developed to treat epilepsy and neuropathic pain but is not believed to act on the same brain receptors. Gabapentin interacts with voltage-gated calcium channels in cortical neurons. It binds to the α2δ subunit (1 and 2) and has been found to reduce calcium currents after chronic but not acute application via an effect on trafficking of voltage-dependent calcium channels in the central nervous system.

Biochem/physiol Actions

Primary Target
α2δ subunit of L-type voltage gated Ca2+ channels

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Eroglu, C. et al. 2009. Cell.139, 380.
Hendrich, J. et al. 2008. Proc. Natl. Acad. Sci. USA105, 3628.
Davies, A. et al. 2007. Trends Pharmacol. Sci.28, 220.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service