Skip to Content
Merck
All Photos(1)

Key Documents

506155

Sigma-Aldrich

Pifithrin-μ

A cell-permeable sulfonamide that blocks p53 interaction with Bcl-xL and Bcl-2 proteins and selectively inhibits p53 translocation to mitochondria without affecting the transactivation function of p53.

Synonym(s):

Pifithrin-μ, Phenylacetylenylsulfonamide, Hsp70 Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NO2S
CAS Number:
Molecular Weight:
181.21
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N)C#Cc1ccccc1

InChI

1S/C8H7NO2S/c9-12(10,11)7-6-8-4-2-1-3-5-8/h1-5H,(H2,9,10,11)

InChI key

ZZUZYEMRHCMVTB-UHFFFAOYSA-N

General description

A cell-permeable sulfonamide that blocks p53 interaction with Bcl-xL and Bcl-2 proteins and selectively inhibits p53 translocation to mitochondria without affecting the transactivation function of p53. Effectively protects against γ radiation-induced cell death in vitro and animal lethality in vivo. Because Pifithrin-μ targets only the mitochondrial branch of the p53 pathway without affecting the important transcriptional functions of p53, it is superior to Pifithrin-α (Cat. No. 506132) in in vivo studies. Shown to selectively interact with inducible HSP70 and disrupt its functions.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
p53 interaction with Bcl-xL and Bcl-2 proteins
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Leu, J., et al. 2009. Mol. Cell36, 15.
Strom, E., et al. 2006. Nat. Chem. Biol.2, 474.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service