Skip to Content
Merck
All Photos(1)

Key Documents

444264

Sigma-Aldrich

MMP Inhibitor III

The MMP Inhibitor III, also referenced under CAS 927827-98-3, controls the biological activity of MMP. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

MMP Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H29N3O4
CAS Number:
Molecular Weight:
363.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥90% (TLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A homophenylalanine-hydroxamic acid based broad-spectrum cell-permeable, reversible inhibitor of matrix metalloproteinases (supplied as a racemic mixture). Inhibits MMP-1 (IC50 = 7.4 nM), MMP-2 (IC50 = 2.3 nM), MMP-3 (IC50 = 135 nM), MMP-7 (IC50 = 10-100 nM), and MMP-13 (IC50 = 1-10 nM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MMP-1
Product does not compete with ATP.
Reversible: yes
Target IC50: 7.4 nM, 2.3 nM, 135 nM, 10-100 nM, 1-10 nM, against MMP-1, MMP-2, MMP-3, MMP-7, and MMP-13, respectively

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months (-20°C).

Other Notes

Covington, M.D., et al. 2006, Am. J. Physiol. Renal. Physiol.290, F43.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Søren B van Witteloostuijn et al.
Journal of peptide science : an official publication of the European Peptide Society, 23(12), 845-854 (2017-10-24)
Bariatric surgery is currently the most effective treatment of obesity, which has spurred an interest in developing pharmaceutical mimetics. It is thought that the marked body weight-lowering effects of bariatric surgery involve stimulated secretion of appetite-regulating gut hormones, including glucagon-like
Elina Siljamäki et al.
Oncogene, 42(49), 3633-3647 (2023-10-21)
Cutaneous squamous cell carcinoma (cSCC) is the most common metastatic skin cancer. The prognosis of patients with metastatic cSCC is poor emphasizing the need for new therapies. We have previously reported that the activation of Ras/MEK/ERK1/2 and transforming growth factor

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service