Skip to Content
Merck
All Photos(1)

Key Documents

344847

Sigma-Aldrich

Fumitremorgin C

from Aspergillus. fumigatus, ≥95% (HPLC), lyophilized, BCRP/ABCG2 inhibitor, Calbiochem®

Synonym(s):

Fumitremorgin C, Aspergillus. fumigatus, NSC719655, FTC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25N3O3
CAS Number:
Molecular Weight:
379.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Fumitremorgin C, Aspergillus. fumigatus, A cell-permeable indolyldiketopiperazinyl mycotoxin that inhibits BCRP/ABCG2 (breast cancer resistance protein/ATP-binding cassette G2) multidrug transport activity.

Quality Level

Assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

solubility

DMSO: 10 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N21[C@@H](Cc4c5c([nH]c4[C@@H]2C=C(C)C)cc(cc5)OC)C(=O)N3[C@@H](CCC3)C1=O

InChI

1S/C22H25N3O3/c1-12(2)9-18-20-15(14-7-6-13(28-3)10-16(14)23-20)11-19-21(26)24-8-4-5-17(24)22(27)25(18)19/h6-7,9-10,17-19,23H,4-5,8,11H2,1-3H3/t17-,18-,19-/m0/s1

InChI key

DBEYVIGIPJSTOR-FHWLQOOXSA-N

General description

A cell-permeable indolyldiketopiperazinyl mycotoxin that inhibits the ATPase and multidrug resistance activity of BCRP/ABCG2 (breast cancer resistance protein/ATP-binding cassette G2) without affecting the activity of MDR1. Also acts as a potent and specific chemosenzitizing agent. Suggested concentration for cell inhibition studies: ~1-5 µM.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
BCRP/ABCG2 (breast cancer resistance protein/ATP-binding cassette G2)
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Allen, J.D., et al. 2002. Mol. Cancer Ther.1, 417.
Ozvegy, C., et al. 2001. Biochem. Biophys. Res. Commun.285, 111.
Rabindran, S.K., et al. 2000. Cancer Res.60, 47.
Rabindran, S.K., et al. 1998. Cancer Res.58, 5850.
Cole, R.J., et al. 1977. J. Agric. Food Chem.25, 826.
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service