Skip to Content
Merck
All Photos(1)

Key Documents

34115-M

Millipore

N-Ethylmaleimide

Sulfhydryl alkylating reagent that inhibits H+-ATPase and suppresses the short circuit current (IC₅₀ = 22 µM) in pancreatic duct cells.

Synonym(s):

N-Ethylmaleimide, NEM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO2
CAS Number:
Molecular Weight:
125.13
MDL number:
UNSPSC Code:
12352111
Pricing and availability is not currently available.

description

Merck USA index - 14, 3822

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

ethanol: 5 mg/mL

storage temp.

10-30°C

SMILES string

N1(C(=O)C=CC1=O)CC

InChI

1S/C6H7NO2/c1-2-7-5(8)3-4-6(7)9/h3-4H,2H2,1H3

InChI key

HDFGOPSGAURCEO-UHFFFAOYSA-N

General description

Sulfhydryl alkylating reagent that inhibits H+-ATPase and suppresses the short circuit current (IC50 = 22 µM) in pancreatic duct cells. Inactivates NADP-dependent isocitrate dehydrogenase. Also a potent inhibitor of both Mg2+ and Ca2+/Mg2+-stimulated DNA fragmentation in rat liver nuclei. Stimulates arachidonic acid release through activation of PLA2 in endothelial cells.
Sulfhydryl alkylating reagent. Has been used for blocking sulfhydryl containing reagents which interfere with glucose oxidase assays. Inactivates NADP-dependent isocitrate dehydrogenase. Endonuclease inhibitor. Also a potent inhibitor of both Mg2+ and Ca2+/Mg2+-stimulated DNA fragmentation in rat liver nuclei.

Biochem/physiol Actions

Primary Target
H+-ATPase
Target IC50: 22 µM in suppressing short circuit current

Warning

Toxicity: Corrosive (L)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Huang, Y.C., et al. 1995. Arch. Biochem. Biophys. 316, 485.
Neve, E.P.A., et al. 1995. Biochem. Pharmacol. 49, 57.
Cain, K., et al. 1994. Biochem. Cell Biol.72, 631.
Banger, K.K., et al. 1993. Biochem. J. 290, 199.
Mattingly, J.R., Jr., et al. 1993. J. Biol. Chem. 268, 3925.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

163.2 °F - closed cup

Flash Point(C)

72.9 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service