Skip to Content
Merck
All Photos(1)

Key Documents

220411

Millipore

Chenodeoxycholic Acid, Sodium Salt

Bile acid that has been shown to increase intracellular Ca2+ in isolated rat hepatocyte couplets. Induces a permeability transition in freshly isolated rat liver mitochondria.

Synonym(s):

Chenodeoxycholic Acid, Sodium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H39O4 · Na
CAS Number:
Molecular Weight:
414.55
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

description

Merck USA index - 14, 2054

Assay

>95% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 20 mg/mL
aqueous buffer: soluble

cation traces

heavy metals: <10 ppm

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Na+].[O-]C(=O)CC[C@H]([C@@H]1[C@@]2([C@H]([C@H]3[C@@H]([C@@]4([C@H](C[C@H]3O)C[C@@H](CC4)O)C)CC2)CC1)C)C

InChI

1S/C24H40O4.Na/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26;/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28);/q;+1/p-1/t14-,15+,16-,17-,18+,19+,20-,22+,23+,24-;/m1./s1

InChI key

WDFRNBJHDMUMBL-OICFXQLMSA-M

General description

Bile acid that has been shown to increase intracellular Ca2+ in isolated rat hepatocyte couplets. Induces a permeability transition in freshly isolated rat liver mitochondria.

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Other Notes

Rolo, A.P., et al. 2001. Biosci. Rep.21, 73.
Thibault, N., and Ballet, F. 1993. Biochem. Pharmacol. 45, 289.
Roda, A., et al. 1990. J. Lipid Res.31, 1433.
Spivak, W., et al. 1988. Biochem. J.252, 275.
Hisadome, T., et al. 1980. Gastroenterol. Jpn.15, 257.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service