Skip to Content
Merck
All Photos(1)

Key Documents

1.94515

Supelco

Acetylacetone

EMPLURA®

Synonym(s):

Acetylacetone, 2,4-Pentanedione

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COCH3
CAS Number:
Molecular Weight:
100.12
Beilstein:
741937
MDL number:
UNSPSC Code:
12352115

vapor density

3.5 (vs air)

vapor pressure

6 mmHg ( 20 °C)

product line

EMPLURA®

autoignition temp.

662 °F

expl. lim.

11.4 %

refractive index

n20/D 1.452 (lit.)

bp

140.4 °C (lit.)

mp

−23 °C (lit.)

density

0.975 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

CC(=O)CC(C)=O

InChI

1S/C5H8O2/c1-4(6)3-5(2)7/h3H2,1-2H3

InChI key

YRKCREAYFQTBPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service