Skip to Content
Merck
All Photos(1)

Documents

1.93410

Supelco

Ascorbic acid

EMPLURA®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352205

product line

EMPLURA®

storage temp.

room temp

Packaging

1.93410.0521: 500 g, Minimum order quantity: 20
1.93410.5021: 5 kg, Minimum order quantity: 4
1.93410.9026: 25 kg, Minimum order quantity: 1

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Inorganics and solvents for basic and standard applications manufactured locally.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service