Skip to Content
Merck
All Photos(3)

Key Documents

17-678

Sigma-Aldrich

ChIPAb+ Trimethyl-Histone H3 (Lys4) - ChIP Validated Antibody and Primer Set

clone CMA304, from mouse, purified by using protein G

Synonym(s):

H3K4me3, Histone H3 (tri methyl K4)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.32

biological source

mouse

Quality Level

clone

CMA304, monoclonal

purified by

using protein G

species reactivity

human, vertebrates

manufacturer/tradename

ChIPAb+
Upstate®

technique(s)

ChIP: suitable (ChIP-seq)
immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction.
The ChIPAb+ Trimethyl-Histone H3 (Lys4) set includes the Anti-trimethyl-Histone H3 (Lys4) antibody, a negative control antibody (purified Mouse IgG), and qPCR primers which amplify a 166 bp region within the promoter of the human GAPDH gene. The trimethyl-Histone H3 (Lys4) and negative control antibodies are supplied in a scalable "per ChIP" reaction size and can be used to functionally validate the precipitation of trimethyl-histone H3 (Lys4)-associated chromatin.
The previously assigned protein identifier Q66I33 has been merged into P84243. Full details can be found on the UniProt database.

Specificity

Recognizes histone H3, Mr 17 kDa, trimethylated at Lys4.
The immunogen sequence is identical in a wide range of animal and plant species.

Immunogen

Epitope: a.a. 1-12
The trimethyl-histone H3 (Lys4) purified antibody is made against a synthetic peptide (trimethylated at Lys4) corresponding to amino acids 1-12 of human histone H3.

Application

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from untreated or colcemid treated HeLa cells (1 X 106 cell equivalents per IP) was subjected to chromatin immunoprecipitation using 2 µg of either a normal mouse IgG or Anti-trimethyl-Histone H3 (Lys4) antibody and the Magna ChIP G Kit (Cat. #17-611). Successful immunoprecipitation of trimethyl-Histone H3 (Lys4) associated DNA fragments was verified by qPCR using primers ampliflying a region of the human B-Globin promoter or using GAPDH promoter Control Primers (Please see figures). Data is presented as percent input of each IP sample relative to input chromatin for each amplicon and ChIP sample as indicated.
Please refer to the EZ-Magna G ChIP (Cat. #17-408) or EZ-ChIP (Cat. #17-371) protocol for experimental details.

Western Blot Analysis:
Representative lot data.
HeLa acid extract were resolved by electrophoresis, transferred to PVDF membrane and probed with Anti-trimethyl Histone H3 (Lys4), clone CMA304 at 1 μg/ml (lane 1) or 0.5 μg/ml (lane 2). Proteins were visualized using a goat anti-mouse secondary antibody conjugated to HRP (Cat. #AP124P) and a chemiluminescence detection system (Please see figures).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
This ChIPAb+ Trimethyl-Histone H3 (Lys4) -ChIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.

Packaging

25 assays per kit, ~2μg per chromatin immunoprecipitation

Quality

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from colcemid-reated HeLa cells (1 X 106 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 2 µg of either normal mouse IgG or Anti-trimethyl-Histone H3 (Lys4) antibody and the Magna ChIP G Kit (Cat. #17-611). Successful immuno-precipitation of trimethyl-histone H3 (Lys4) associated DNA fragments was verified by qPCR using Control Primers (Please see figures).
Please refer to the EZ-Magna G ChIP (Cat. #17-409) or EZ-ChIP (Cat. #17-371) protocol for experimental details.

Target description

Trimethyl histone H3 at ~17 kDa

Physical form

Anti-trimethyl-Histone H3 (Lys4) (mouse monoclonal IgG1, Clone CMA304). One vial containing 50 μg of protein G purified antibody in 50 μL PBS containing 0.05% sodium. Store at -20°C.

Normal Mouse IgG. Two vials containing 25 μg purified mouse IgG in 25 μL storage buffer containing 0.1% sodium azide. Store at -20°C.

Control Primers. One vial containing 75 μL of 5 μM of each primer specific for the promoter region of GAPDH. Store at -20°C.
FOR: TAC TAG CGG TTT TAC GGG CG
REV: TCG AAC AGG AGG AGC AGA GAG CGA

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Aliquot upon thawing, avoid freeze thaw cycles.

Analysis Note

Control
Included negative control mouse IgG antibody and control primers specific for human GAPDH promoter.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The gene signature in CCAAT-enhancer-binding protein ? dysfunctional acute myeloid leukemia predicts responsiveness to histone deacetylase inhibitors.
Liss, A; Ooi, CH; Zjablovskaja, P; Benoukraf, T; Radomska, HS; Ju, C; Wu, M; Balastik et al.
Haematologica null
Pei Wang et al.
Drug metabolism and disposition: the biological fate of chemicals, 49(5), 361-368 (2021-03-07)
The maintenance of homeostasis of cytochromes P450 enzymes (P450s) under both physiologic and xenobiotic exposure conditions is ensured by the action of positive and negative regulators. In the current study, the hepatocyte nuclear factor 4α (HNF4A) antisense RNA 1 (HNF4A-AS1)
Jonathan C Irish et al.
Molecular oncology, 10(6), 850-865 (2016-03-24)
The 8p11-p12 amplicon occurs in approximately 15% of breast cancers in aggressive luminal B-type tumors. Previously, we identified WHSC1L1 as a driving oncogene from this region. Here, we demonstrate that over-expression of WHSC1L1 is linked to over-expression of ERα in
Weiping Wang et al.
Molecular and cellular endocrinology, 428, 38-48 (2016-03-21)
The mechanism of insulin gene transcription control in response to glucose concentration is poorly defined. The islet-restricted transcription factors PDX1 and ISL1 interact with BETA2, activating insulin gene expression. However, their contribution and hierarchical organization in insulin expression control based
Jie Wu et al.
Immunity, 47(6), 1114-1128 (2017-12-10)
CD4+ T cells orchestrate immune responses and destruction of allogeneic organ transplants, but how this process is regulated on a transcriptional level remains unclear. Here, we demonstrated that interferon regulatory factor 4 (IRF4) was a key transcriptional determinant controlling T cell responses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service