Skip to Content
Merck

1.37162

SAFC

D(-)-Mannitol

EMPROVE® EXPERT, Ph. Eur., BP, ChP, JP, USP

Pharma Manufacturing

Synonym(s):

D-Mannitol, Mannite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721898
MDL number:
UNSPSC Code:
12352201
Pricing and availability is not currently available.

Agency

BP
ChP
JP
Ph. Eur.
USP

Quality Level

product line

EMPROVE® EXPERT

form

solid

impurities

low in endotoxins

pH

5.0-6.5 (120 °C, 182 g/L)

bp

290-295 °C/4.67 hPa

mp

166-169 °C
167-170 °C (lit.)

solubility

213 g/L

application(s)

liquid formulation
ophthalmics
parenterals
pharma/biopharma processes

storage temp.

2-25°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-KVTDHHQDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemicals play an important role in the stabilization of a biologic drug during its manufacturing and formulation process – for instance, by preventing aggregation. We offer a wide range of high-quality stabilizers, buffers and salts to successfully purify and formulate your biomolecules. Specifically developed for high-risk applications, they are low in bioburden and endotoxins. As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Mannitol is a sugar alcohol which is used widely in biomolecule formulations as a stabilizer, bulking agent, and lyoprotecting agent, especially in lyophilization. Due to its low microbial and endotoxin limits, D(-)-Mannitol Emprove® Expert is especially suitable for high-risk applications.

Biochem/physiol Actions

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Learn more on mAb subcutaneous or intravenous formulation, and the relevant associated products, including excipients.

Learn more on mAb subcutaneous or intravenous formulation, and the relevant associated products, including excipients.

Learn more on mAb subcutaneous or intravenous formulation, and the relevant associated products, including excipients.

Learn more on mAb subcutaneous or intravenous formulation, and the relevant associated products, including excipients.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service