Skip to Content
Merck
All Photos(1)

Key Documents

12-523

Sigma-Aldrich

GST, 6 His-tagged

GST, 6 His-tagged

Synonym(s):

GST Fusion Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
eCl@ss:
32160405
NACRES:
NA.41
Pricing and availability is not currently available.

form

liquid

Quality Level

manufacturer/tradename

Upstate®

technique(s)

affinity chromatography: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Glutathione S-transferase (GST) belongs to three families of proteins distinguished as cytosolic, mitochondrial, and microsomal GST. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: α, κ, μ, ω, π, σ, θ, and ζ. This product is a C-terminal 6 histidine-tagged, recombinant full-length glutathione S-transferase (GST).

Biochem/physiol Actions

Glutathione S-transferases (GSTs) are a family of enzymes that play an important role in phase II detoxification by catalyzing the conjugation of many hydrophobic and electrophilic compounds with reduced glutathione. The glutathione S-transferase pi gene (GSTP1) is a polymorphic gene encoding active, functionally different GSTP1 variant proteins that are thought to function in xenobiotic metabolism and play a role in susceptibility to cancer, and other diseases.

Physical form

50mM Tris-HCl, pH 8, 150mM NaCl, 20% glycerol

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service