Skip to Content
Merck
All Photos(1)

Documents

1.11277

Millipore

m-ENDO agar LES

GranuCult® plus, APHA, suitable for microbiology, for coliforms

Synonym(s):

Coliform selective agar, modified Endo medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

APHA

Quality Level

sterility

non-sterile

form

granular

packaging

pkg of 500 g

manufacturer/tradename

GranuCult® plus

storage condition

(Tightly closed. Dry. Keep in a well-ventilated place.)

technique(s)

microbiological culture: suitable

color

dark violet

pH

7.0-7.4 (25 °C in H2O, ready-to-use solution; after autoclaving)

solubility

51 g/L

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

coliforms

General description

m-ENDO Agar LES (Lawrence Experimental Station) is a medium recommended for the enumeration of coliforms in water used in the Standard Total Coliform Membrane Filter Procedure. The American Public Health Association (APHA) recommends m-Endo Agar in standard total coliform membrane filtration for testing potable water and wastewater. It follows the two-step membrane filter procedure using Lauryl Sulfate Broth as a preliminary enrichment, resulting in higher coliform counts. Peptones in the medium are a source of carbon, nitrogen, vitamins, and minerals. Yeast extract supplies B-complex vitamins, which stimulate bacterial growth. Lactose is a carbohydrate. Phosphates are buffering agents. Sodium chloride maintains the osmotic balance of the medium. Sodium desoxycholate and sodium lauryl sulfate are added to inhibit the growth of the accompanying flora. Pararosaniline (fuchsin) is an indicator. Sodium sulfite is added to decolorize the basic fuchsin solution. Agar is the solidifying agent. Lactose-positive colonies are colored red due to the liberation of fuchsin from the fuchsin-sulfite compound. E. coli colonies develop a metallic sheen due to the production of aldehydes with the rapid fermentation of lactose. Lactose-nonfermenting bacteria appear as clear, colorless colonies.

Application

m-ENDO agar LES is suitable for the enumeration of coliforms in water using a two-step membrane filtration technique.

Features and Benefits

  • GranuCult® offers superior granulated culture media
  • Safe and sustainable due to reduced risks associated with fine dust and toxic substance inhalation, resulting in a safer work environment
  • Excellent wettability, solubility, and free-flowing properties
  • Convenient, with minimal component separation and clumping, even under warm or humid conditions
  • High batch-to-batch reproducibility
  • Prolonged shelf life of up to five years
  • High number of test strains exceeding all regulatory demands
  • Granulation technology allows many supplements to be included, with no need to add these separately

Analysis Note

Appearance (clearness): clear to turbid
Appearance (color): red
pH-value (25 °C): 7.0 - 7.4
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): good to very good
Growth (Enterobacter aerogenes ATCC 13048): good to very good
Growth (Proteus mirabilis ATCC 14273): good to very good
Growth (Staphylococcus aureus ATCC 25923 (WDCM 00034)): none
Growth (Enterococcus faecalis ATCC 19433 (WDCM 00009)): none to poor
Colony color (Escherichia coli ATCC 25922 (WDCM 00013)): red
Colony color (Enterobacter aerogenes ATCC 13048): red
Colony color (Proteus mirabilis ATCC 14273): reddish/red
Metallic luster (Escherichia coli ATCC 25922 (WDCM 00013)): +
Metallic luster (Enterobacter aerogenes ATCC 13048): +
Metallic luster (Proteus mirabilis ATCC 14273): -
Incubation: 24 hrs.; 35 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service