Skip to Content
Merck
All Photos(1)

Key Documents

110165CBC

Sigma-Aldrich

Thiamet G

TMG

Synonym(s):

O-GlcNAcase Inhibitor, (3aR,5R,6S,7R,7aR)-2-(ethylamino)-3a,6,7,7a-tetrahydro-5-(hydroxymethyl)-5H-Pyrano[3,2-d]thiazole-6,7-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16N2O4S
CAS Number:
Molecular Weight:
248.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

SMILES string

CCNC1=N[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@@H]2S1

InChI

1S/C9H16N2O4S/c1-2-10-9-11-5-7(14)6(13)4(3-12)15-8(5)16-9/h4-8,12-14H,2-3H2,1H3,(H,10,11)/t4-,5-,6-,7-,8-/m1/s1

InChI key

PPAIMZHKIXDJRN-FMDGEEDCSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Thiamet G is a potent inhibitor of the enzyme O-GlcNAcase (Ki = 21 nM). The compound is orally bioavailable and crosses the blood brain barrier. Thiamet G leads to an increase in O-GlcNAc-modified proteins in cell-based and in vivo assay systems, and reduces levels of phosphorylated Tau protein in rat cortex and hippocampus.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service