1.09174
MAS-100 Iso NT® Remote Microbial Air Sampler
with USB, digital I/O and Ethernet interfaces
Synonym(s):
MAS-100 Iso NT® Control unit
About This Item
Recommended Products
material
aluminum hardware
Agency
according to ISO 14698
feature
installed
packaging
pkg of 1 kit
manufacturer/tradename
MAS-100 Iso NT®
application(s)
air monitoring
pharmaceutical
compatibility
for use with 90 mm Petri dishes
storage temp.
no temp limit
General description
The innovative double valve system enables the sampling heads to be integrated into the decontamination process with VHP. Each unit has both an air and a sterilization pump that operate autonomously. The instrument can be operated via an LCD screen.
The fully automatic calibration of the device guarantees maximum efficiency and accuracy. The system measures the air flow and regulates it to a constant value of 100 liters/min. These systems have the highest airflow accuracy of ±2.5%, compared to others that can be as high as ±10%. The instruments are designed and built according to GAMP 4 and are independently validated according to ISO 14698-1 with very good physical efficiency providing a D50 value below 1.1μm and a high biological efficiency result. The efficiencies and the D50 values are independent of the tube length and are comparable to the other members of the MAS-family.
The MAS-100 ISO NT® is equipped with one valve for one sampling head and RS232, USB and digital IN/OUT interfaces for communication. In case another communication interface is required MAS-100 ISO NT® Control unit with Profibus with Profibus interface is available. The type of sample base (standard or easy clean version) as well as the perforated lids can be selected separately according to the specific needs of the sample location and workflow.
Application
Components
Legal Information
accessory
related product
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service