1.08483
Urea broth
for detecting microorganisms which metabolize urea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106213
Recommended Products
Quality Level
form
powder (dehydrated (DCM))
manufacturer/tradename
GranuCult®
technique(s)
microbiological culture: suitable
pH
6.7-6.9 (37 °C, 39 g/L in H2O, after autoclaving)
solubility
39 g/L
bulk density
810 kg/m3
application(s)
clinical
microbiology
storage temp.
15-25°C
suitability
Proteus spp.
General description
for detecting microorganisms which metabolize urea
Analysis Note
Appearance (clearness)clearAppearance (colour)orange-redpH-value (25 °C)6.6 - 7.0colour change to red (Escherichia coli ATCC 25922 (WDCM 00013))-colour change to red (Salmonella typhimurium ATCC 14028 (WDCM 00031))-colour change to red (Klebsiella pneumoniae ATCC 13883 (WDCM 00097))-colour change to red (Proteus hauseri ATCC 13315)+colour change to red (Proteus mirabilis ATCC 14153)+colour change to red (Proteus rettgeri ATCC 29944)+Incubation: 48 hrs.; 35 °C; aerobic.
Footnote
We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service