Skip to Content
Merck
All Photos(1)

Key Documents

1.07372

Sigma-Aldrich

Lead(II) acetate trihydrate

extra pure

Synonym(s):

Bis(acetato)trihydroxytrilead, Lead acetate trihydrate, Lead diacetate trihydrate, Plumbous acetate trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(CH3CO2)2 · 3H2O
CAS Number:
Molecular Weight:
379.33
Beilstein:
3730298
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

form

solid

pH

5.5-6.5 (20 °C, 50 g/L in H2O)

mp

75 °C (dec.) (lit.)

density

3.3 g/cm3 at 20 °C

SMILES string

O.O.O.CC(=O)O[PbH2]OC(C)=O

InChI

1S/2C2H4O2.3H2O.Pb/c2*1-2(3)4;;;;/h2*1H3,(H,3,4);3*1H2;/q;;;;;+2/p-2

InChI key

MCEUZMYFCCOOQO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Lead(II) acetate trihydrate can be used:
  • As a lead precursor to fabricate large area absorber layers of planar inverted perovskite solar cells by blade coating technique.[1]
  • To modify lightweight graphite felt electrodes as current collectors applicable in lead-acid batteries.[2]
  • As a starting material to prepare perovskite lead magnesium niobate (PMN) powder applicable in the manufacturing of actuators, multilayer ceramic capacitors, and electro-optic devices.[3]
  • To prepare PbS quantum dots applicable in the fabrication of hybrid-organic photodetector for X-ray detection.[4]

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1A - STOT RE 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modification of Graphite Felt with Lead (II) Formate and Acetate?An Approach for Preparation of Lightweight Electrodes for a Lead-Acid Battery
Ilginis A and Griskonis E
Processes, 8(10), 1248-1248 (2020)
Phase formation and microstructure of Nd+3 doped Pb(Mg1/3Nb2/3)O3 prepared by sol?gel method
Aylin Sakar-Deliormanli A, et al.
Journal of Materials Science: Materials in Electronics, 19, 577-583 (2008)
PbS quantum dot based hybrid-organic photodetectors for X-ray sensing
Author links open overlay panel
Ankah GN, et al.
Organic Electronics, 33, 201-206 (2016)
A Matter of Drying: Blade-Coating of Lead Acetate Sourced Planar Inverted Perovskite Solar Cells on Active Areas >1cm2
Kohlstadt M, et al.
Physica Status Solidi, 215(21), 1800419-1800419 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service