Skip to Content
Merck

1.06360

SAFC

Sodium bromide

EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, USP

Pharma Manufacturing

Synonym(s):

Sodium bromide, Bromo sodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BrNa
CAS Number:
Molecular Weight:
102.89
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-599-9

Agency

BP
JP
Ph. Eur.
USP

Quality Level

vapor pressure

1.3 hPa ( 806 °C)

product line

EMPROVE® ESSENTIAL

form

solid

technique(s)

API processing | salt formation: suitable

pH

5.74 (22.5 °C, 430 g/L in H2O)

solubility

946 g/L

density

3.2 g/cm3 at 25 °C

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

15-25°C

SMILES string

[Na+].[Br-]

InChI

1S/BrH.Na/h1H;/q;+1/p-1

InChI key

JHJLBTNAGRQEKS-UHFFFAOYSA-M

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sodium bromide is a mineral salt with diverse pharmaceutical applications, especially in veterinary medicine.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service