Skip to Content
Merck
All Photos(1)

Key Documents

106313-M

SAFC

CellPrime® rTransferrin

recombinant, expressed in yeast, from synthetic

Pharma Manufacturing

Synonym(s):

CellPrime® rTransferrin, transferrin, transferrins

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
80000
MDL number:
UNSPSC Code:
41106213
EC Index Number:
234-318-8

biological source

synthetic

Quality Level

recombinant

expressed in yeast

description

recombiant human transferrin

sterility

sterile (low bioburden)

parameter

2-8 °C temp. range

storage temp.

2-8°C

General description

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.
recombinant human transferrin, expressed in Pichia pastoris

Application

media manufacturing supplement

Features and Benefits

safe and suitable NAO recombinant supplement

Packaging

  • 1063130010: 10 ml ( 2%, w/v)
  • 1063130050: 50 ml ( 2%, w/v)

Shipped in blue ice packs.

Other Notes

Product is manufactured in a facility adhering to Good Manufacturing Practices.

Caution

do not freeze

Legal Information

CellPrime is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service