Skip to Content
Merck
All Photos(1)

Key Documents

1.06056

Supelco

2-Methylbutane

for spectroscopy Uvasol®

Synonym(s):

2-Methylbutane, iso-Pentane, Isopentane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12
CAS Number:
Molecular Weight:
72.15
MDL number:
UNSPSC Code:
12191502
EC Index Number:
201-142-8
NACRES:
NA.03
Assay:
≥99.8% (GC)
technique(s):
UV/Vis spectroscopy: suitable
bp:
28 °C/1013 hPa
vapor pressure:
800 hPa ( 20 °C)
Pricing and availability is not currently available.

Agency

reag. Ph. Eur.

Quality Level

vapor pressure

800 hPa ( 20 °C)

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

420 °C

expl. lim.

1.3-7.6 % (v/v)

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.005% Water

evapn. residue

≤0.0005%

color

APHA: ≤10

transmittance

190 nm, ≥50%
200 nm, ≥65%
210 nm, ≥85%
215 nm, ≥90%
240 nm, ≥98%

bp

28 °C/1013 hPa

mp

-160 °C

transition temp

flash point -57 °C

density

0.62 g/cm3 at 20 °C

UV absorption

λ: 190 nm Amax: ≤0.30
λ: 200 nm Amax: ≤0.19
λ: 210 nm Amax: ≤0.07
λ: 215 nm Amax: ≤0.05
λ: 240 nm Amax: ≤0.01

storage temp.

2-30°C

SMILES string

C(CC)(C)C

InChI

1S/C5H12/c1-4-5(2)3/h5H,4H2,1-3H3

InChI key

QWTDNUCVQCZILF-UHFFFAOYSA-N

General description

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol<TMSYMBOL></TMSYMBOL> solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol<TMSYMBOL></TMSYMBOL> solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified.

Analysis Note

Purity (GC): ≥ 99.8 %
Evaporation residue: ≤ 0.0005 %
Water: ≤ 0.005 %
Colour: ≤ 10 Hazen
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Fluorescence (as quinine at 254 nm): ≤ 1.0 ppb
Fluorescence (as quinine at 365 nm): ≤ 1.0 ppb
Transmission (at 190 nm): ≥ 50 %
Transmission (at 200 nm): ≥ 65 %
Transmission (at 210 nm): ≥ 85 %
Transmission (at 215 nm): ≥ 90 %
Transmission (from 240 nm): ≥ 98 %
Absorbance (at 190 nm): ≤ 0.30
Absorbance (at 200 nm): ≤ 0.19
Absorbance (at 210 nm): ≤ 0.07
Absorbance (at 215 nm): ≤ 0.05
Absorbance (from 240 nm): ≤ 0.01
Meet the requirements of "Specific use spectroscopy" according Reag. Ph Eur

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

UVASOL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-59.8 °F - closed cup

Flash Point(C)

-51 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. what type of Cap is on these flasks

    1 answer
    1. This product is packaged in a glass bottle with a polytetrafluoroethylene (PTFE) cap.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service