Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
1.04417
Mercury(II) chloride
fine cryst. EMPLURA®
Synonym(s):
Mercury(II) chloride, Mercury dichloride
About This Item
Recommended Products
vapor pressure
0.0001 hPa ( 20 °C)
Quality Level
product line
EMPLURA®
Assay
99.5-100.5% dry basis (complexometric)
form
solid
loss
≤ 1.0% loss on drying
pH
3.2 (15 g/L in H2O)
bp
302 °C/1013 hPa (sublimed)
mp
280.7 °C
density
5.44 g/cm3 at 20 °C
bulk density
2000 kg/m3
cation traces
HgCl2:, passes test
storage temp.
2-30°C
SMILES string
[Hg+2].[Cl-].[Cl-]
InChI
1S/2ClH.Hg/h2*1H;/q;;+2/p-2
InChI key
LWJROJCJINYWOX-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Application
- Unveiling the Dual Nature of Heavy Metals: Stressors and Promoters of Phenolic Compound Biosynthesis in Basilicum polystachyon (L.) Moench In Vitro.: This study investigates the role of Mercury(II) chloride in modulating phenolic compound biosynthesis, demonstrating its dual nature as both a stressor and a promoter in plant in vitro systems (Das et al., 2023).
- Development of methodology to generate, measure, and characterize the chemical composition of oxidized mercury nanoparticles.: This research develops novel analytical methods for the generation and characterization of oxidized mercury nanoparticles using Mercury(II) chloride, contributing to advancements in environmental monitoring and analytical chemistry (Ghoshdastidar et al., 2020).
- A Study of the Complexation of Mercury(II) with Dicysteinyl Tetrapeptides by Electrospray Ionization Mass Spectrometry.: This paper explores the complexation of Mercury(II) chloride with peptides, enhancing understanding of its interactions and potential analytical applications in biochemistry (Mazlo and Ngu-Schwemlein, 2016).
- Quantitative Bioimaging to Investigate the Uptake of Mercury Species in Drosophila melanogaster.: The study uses advanced quantitative bioimaging techniques to examine how Drosophila melanogaster absorbs different mercury species, including those derived from Mercury(II) chloride, highlighting its potential for environmental impact studies (Niehoff et al., 2015).
Analysis Note
Identity: passes test
Appearance of solution: passes test
Acidity or alkalinity: passes test
Mercury(I)-chloride: passes test
Loss on drying: ≤ 1.0 %
Legal Information
Disclaimer

Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 1
Storage Class Code
6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
-
Is it can be used in ayurvedic medicine
1 answer-
Unless otherwise stated this product is intended for research use only and is not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service