Skip to Content
Merck
All Photos(1)

Key Documents

1.01220

Supelco

Ammonium amidosulfonate

for analysis (for detection of sulfonamide in blood) EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Ammonium amidosulfonate, Amidosulfonic acid ammonium salt, Ammonium sulfamate, Sulfamic acid ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H6N2O3S
CAS Number:
Molecular Weight:
114.12
MDL number:
UNSPSC Code:
12352301
EC Index Number:
231-871-7
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

≥99.0% (acidimetric)

form

solid

potency

2000 mg/kg LD50, oral (Rat)

impurities

≤0.02% In water insoluble matter

ign. residue

≤0.05% (as SO4)

loss

≤0.5% loss on drying, 105°C

pH

5.0-6.0 (20 °C, 50 g/L in H2O)

bp

160 °C/1013 hPa (decomposes)

mp

132-134 °C

bulk density

680 kg/m3

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.2%

cation traces

Fe: ≤0.0005%
heavy metals: ≤0.0005%

storage temp.

no temp limit

SMILES string

[S](=O)(=O)(N)O.N

InChI

1S/H3NO3S.H3N/c1-5(2,3)4;/h(H3,1,2,3,4);1H3

InChI key

GEHMBYLTCISYNY-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 99.0 %
In water insoluble matter: ≤ 0.02 %
Melting point: 132.0 - 134.0 °C
pH-value (5 %; water): 4.5 - 5.5
Chloride (Cl): ≤ 0.05 %
Sulphate (SO₄): ≤ 0.2 %
Heavy metals: ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
Residue on ignition (as SO₄): ≤ 0.05 %
Loss on Drying (105°C): ≤ 0.5 %
ACS and Reag. Ph Eur conforms

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service