Skip to Content
Merck
All Photos(2)

Key Documents

1.00888

Millipore

Clostridium perfringens selective supplement

For the preparation of TSC Agar, For the enumeration of Clostridium perfringens

Synonym(s):

Clostridium perfringens Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106212
NACRES:
NA.74
Pricing and availability is not currently available.

Quality Level

form

lyophilized powder

technique(s)

microbiological culture: suitable

solubility

83 g/L

application(s)

food and beverages
microbiology

storage temp.

2-8°C

suitability

Clostridium spp.

General description

Clostridium perfringens selective supplement contains the antibiotic D-Cycloserine that inhibits the accompanying bacterial flora and causes the developing colonies to remain smaller. It also reduces the diffusion and the disturbing blackening around the Clostridium perfringens colonies. 4-Methylumbelliferylphosphate (MUP) is a fluorogenic substrate for the alkaline and acid phosphatase. The acid phosphatase is a high specific indicator for Clostridium perfringens, and it splits the fluorogenic substrate MUP. The fluorescence of resulting product, 4-methylumbelliferone, can be identified in long wave UV-light, which suggests the presence of Clostridium perfringens.

Application

Clostridium perfringens selective supplement is used for the enrichment of TSC-Agar (Base) during the enumeration of Clostridium perfringens in food products and other materials.

Linkage

Replaces: 1008880017

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service