Skip to Content
Merck
All Photos(1)

Key Documents

1.00380

Sigma-Aldrich

ISOSLIDE Iron Control Slides

with reference tissue for the detection of free iron in histological tissue

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124
NACRES:
NA.41
Pricing and availability is not currently available.

Quality Level

IVD

for in vitro diagnostic use

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

The ISOSLIDE Iron Control Slides - with reference tissue for the detection of free iron in histological tissue, are used for human-medical cell diagnosis and serve the purpose of the histological investigation and quality control of sample material of human origin. They are ready-to-use glass slides with paraffin sections that contain the typical structures of the corresponding test method.
The ISOSLIDE Iron Control Slides are prepared using sections of suitable animal material, that clearly visualize ionic iron (Fe3+) not bound to the heme structure. Each package of ISOSLIDE control slides contains 25 slides. The tissue is applied onto the slide next to the white printed labelling field. One slide is already stained with the reference method (HEMATOGNOST Fe - staining kit, Product number. 1.12084) and is used for comparison. The 24 unstained slides are stained according to the corresponding protocol or with the laboratory′s own protocol. After the staining of the control slides, the staining result is to be compared with the laboratory material and the pre-stained control slide. For more details, please see instructions for use (IFU). This product is registered as IVD and CE marked. The IFU can be downloaded from this webpage.

Application

Control slides with reference tissue for the detection of free iron in histological tissue

Analysis Note

Suitability for microscopypasses testIronIron detectable

Legal Information

HEMATOGNOST FE is a trademark of Merck KGaA, Darmstadt, Germany
ISOSLIDE is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service