Skip to Content
Merck
All Photos(3)

Key Documents

07-2162

Sigma-Aldrich

Anti-PCNA Antibody

from rabbit

Synonym(s):

proliferating cell nuclear antigen, cyclin, DNA polymerase delta auxiliary protein, Cyclin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse, human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PCNA(5111)

General description

Proliferating cell nuclear antigen (PCNA) belongs to the DNA sliding clamp family and is comprised of three identical monomers forming a heterodimer. Due to its unique structure, PCNA is able to increase the processivity of DNA polymerase delta by using its outer domains to be involved in protein-protein interaction while still tethered to and freely sliding along the DNA. PCNA is a critical regulatory component in the onset of leading strand DNA replication and the termination of lagging strand synthesis. Studies have also indicated a role for PCNA in the recruitment and regulation of MMR pathway repair proteins. PCNA is also able to interact with a wide variety of cell cycle proteins and serves to coordinate several proteins participating in DNA processes, such as apoptosis.

Specificity

Other homologies: Bovine, rabbit, canine, and panda (99% sequence homology).

Immunogen

GST-tagged recombinant protein corresponding to human PCNA.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Use Anti-PCNA Antibody (Rabbit Polyclonal Antibody) validated in WB, ICC to detect PCNA also known as proliferating cell nuclear antigen, cyclin, DNA polymerase delta auxiliary protein, Cyclin.
Western Blot Analysis: 1 µg/mL from a previous lot detected PCNA on 10 µg of NIH/3T3 and PC12 cell lysates.

Immunocytochemistry Analysis: 1:500 dilution from a previous lot detected PCNA in A431 cells.

Quality

Evaluated by Western Blot in A431 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected PCNA on 10 µg of A431 cell lysate.

Target description

~ 35 kDa observed. PCNA has a predicted molecular weight of 29 kDa, however, it can be monoubiquitinated, resulting in a ~ 35 kDa band.

Physical form

Format: Purified
Protein A
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A431 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service