05-852
Anti-M-Cadherin Antibody, clone 12G4
clone 12G4, Upstate®, from mouse
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
12G4, monoclonal
species reactivity
mouse
manufacturer/tradename
Upstate®
technique(s)
ELISA: suitable
immunocytochemistry: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
mouse ... Cdh15(12555)
Specificity
M-Cadherin
Immunogen
Recombinant extracellular domain of mouse M-Cadherin
Application
Anti-M-Cadherin Antibody, clone 12G4 is an antibody against M-Cadherin for use in ELISA, WB & IC.
Research Category
Cell Structure
Cell Structure
Research Sub Category
Adhesion (CAMs)
Adhesion (CAMs)
Quality
routinely evaluated by immunoblot on modified RIPA lysates from C2C12 cells
Target description
Mr 130kDa
Physical form
Format: Purified
Thiophilic adsorption and size exclusion
Storage and Stability
Stable for 1 year at -20°C from date of shipment
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Cell and tissue research, 296(1), 191-198 (1999-04-13)
Cadherins are calcium-dependent, transmembrane intercellular adhesion proteins with morphoregulatory functions in the development and maintenance of tissues. In the development of striated muscle, the expression and function of mainly M-, N-, and R-cadherin has been studied so far. While these
Convergence of Wnt, beta-catenin, and cadherin pathways.
Science (New York, N.Y.), 303, 1483-1487 (2004)
The Journal of biological chemistry, 273(16), 10011-10018 (1998-05-23)
We used a novel cDNA cloning method based on the cadherin-beta-catenin protein interaction and identified a new human classic-type cadherin, which we named cadherin-15, from adult brain and skeletal muscle cDNA libraries. Sequence analysis revealed that this cadherin was closely
Structure-based models of cadherin-mediated cell adhesion: the evolution continues.
Cellular and Molecular Life Sciences, 61, 1884-1895 (2004)
The cadherin-catenin complex as a focal point of cell adhesion and signalling: new insights from three-dimensional structures.
Bioessays, 26, 497-511 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service