Skip to Content
Merck
All Photos(1)

Documents

04-1469

Sigma-Aldrich

Anti-hnRNP A1 Antibody, clone 9H10

clone 9H10, from mouse

Synonym(s):

Helix-destabilizing protein, Single-strand RNA-binding protein, heterogeneous nuclear ribonucleoprotein A1, heterogeneous nuclear ribonucleoprotein A1B protein, heterogeneous nuclear ribonucleoprotein B2 protein, heterogeneous nuclear ribonucleoprotein c

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9H10, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... HNRNPA1(3178)

General description

The hnRNPs are RNA binding proteins that complex with heterogeneous nuclear RNA (hnRNA) and associate with pre-mRNAs in the nucleus. These complexes are associated with pre-mRNA processing and other aspects of mRNA metabolism and transport. While all hnRNPs are present in the nucleus, data suggests they shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by hnRNP A1 has two repeats of quasi-RRM domains that bind to RNAs. This abundant core protein, along with other hnRNP proteins, is exported from the nucleus, probably bound to mRNA, and is immediately re-imported. The hnRNP A1 protein is involved in the packaging of pre-mRNA into hnRNP particles, transport of poly A+ mRNA from the nucleus to the cytoplasm, and may have a role in splice site selection.

Specificity

This antibody recognizes hnRNP A1.

Immunogen

Epitope: Unknown
Recombinant protein corresponding to human hnRNP A1.

Application

Anti-hnRNP A1 Antibody, clone 9H10 is a Mouse Monoclonal Antibody for detection of hnRNP A1 also known as Helix-destabilizing protein or Single-strand RNA-binding protein & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins

Quality

Evaluated by Western Blot in HeLa nuclear extract.

Western Blot Analysis: 0.01 µg/ml of this antibody detected hnRNP A1 on 10 µg of HeLa nuclear extract.

Target description

~ 38 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa nuclear extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chung-Te Chang et al.
The EMBO journal, 32(3), 473-486 (2013-01-10)
The TREX complex couples nuclear pre-mRNA processing with mRNA export and contains multiple protein components, including Uap56, Alyref, Cip29 and the multi-subunit THO complex. Here, we have identified Chtop as a novel TREX component. We show that both Chtop and
Inhibition of vascular endothelial growth factor receptor 2-mediated endothelial cell activation by Axl tyrosine kinase receptor.
Margherita Gallicchio,Stefania Mitola,Donatella Valdembri,Roberto Fantozzi,Brian Varnum et al.
Blood null
Yunji Zheng et al.
Journal of virology, 94(20) (2020-08-08)
Human papillomavirus 16 (HPV16) 5'-splice site SD226 and 3'-splice site SA409 are required for production of the HPV16 E7 mRNAs, whereas unspliced mRNAs produce E6 mRNAs. The E6 and E7 proteins are essential in the HPV16 replication cycle but are
Joshua N Douglas et al.
Journal of neuroinflammation, 13(1), 178-178 (2016-07-09)
Neurodegeneration is believed to be the primary cause of permanent, long-term disability in patients with multiple sclerosis. The cause of neurodegeneration in multiple sclerosis appears to be multifactorial. One mechanism that has been implicated in the pathogenesis of neurodegeneration in
Joshua N Douglas et al.
Journal of clinical & cellular immunology, 7(2), 402-402 (2016-07-05)
Multiple sclerosis (MS) is the most common demyelinating disorder of the central nervous system (CNS). Data suggest that antibodies to CNS targets contribute to the pathogenesis of MS. MS patients produce autoantibodies to heterogeneous nuclear ribonucleoprotein A1 (hnRNP A1). hnRNP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service