Skip to Content
Merck
All Photos(1)

Key Documents

Z-007

Supelco

Zolpidem phenyl-4-carboxylic acid solution

500 μg/mL in acetonitrile: water (1:1), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

500 μg/mL in acetonitrile: water (1:1)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CN(C)C(=O)Cc1c(nc2ccc(C)cn12)-c3ccc(cc3)C(O)=O

InChI

1S/C19H19N3O3/c1-12-4-9-16-20-18(13-5-7-14(8-6-13)19(24)25)15(22(16)11-12)10-17(23)21(2)3/h4-9,11H,10H2,1-3H3,(H,24,25)

InChI key

FELZONDEFBLTSP-UHFFFAOYSA-N

General description

Zolpidem Phenyl-4-carboxylic acid is the major urinary metabolite of zolpidem, accounting for 51% of an administered dose. Literature references indicate the metabolite can be found in urine for 2-3 days following ingestion of a single therapeutic dose of zolpidem.[1] Zolpidem, marketed as Ambien® or Stilnox®, is a nonbenzodiazepine used for the short-term treatment of insomnia.

Legal Information

Ambien is a registered trademark of Sanofi S.A.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Stilnox is a registered trademark of Sanofi S.A.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John H Lewis et al.
Journal of analytical toxicology, 31(4), 195-199 (2007-06-09)
Zolpidem is a non-benzodiazepine hypnotic that has been implicated in both drug-facilitated sexual assault and drink spiking. Detection of the drug in urine is extremely difficult because of its extensive metabolism. A method is presented for the detection and quantitation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service