Skip to Content
Merck
All Photos(1)

Key Documents

S-043

Supelco

JWH-019 6-Hydroxyhexyl metabolite solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

JWH-019 6-Hydroxyhexyl metabolite solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H25NO2
CAS Number:
Molecular Weight:
371.47
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

OCCCCCCn1cc(C(=O)c2cccc3ccccc23)c4ccccc14

InChI

1S/C25H25NO2/c27-17-8-2-1-7-16-26-18-23(21-13-5-6-15-24(21)26)25(28)22-14-9-11-19-10-3-4-12-20(19)22/h3-6,9-15,18,27H,1-2,7-8,16-17H2

InChI key

LBVZVDUWOZUDJT-UHFFFAOYSA-N

General description

JWH-019 6-Hydroxyhexyl metabolite is a major urinary metabolite of JWH-019, a synthetic cannabinoid found in several blends of the herbal mixture Spice or K2. This biomarker is also referred to as the omega hydroxy metabolite of JWH-019 since the hydroxy group is located at the terminal position of the N-hexyl side chain.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service