Skip to Content
Merck
All Photos(1)

Key Documents

P-122

Supelco

Prednisone solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O5
CAS Number:
Molecular Weight:
358.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

-48 °C

density

0.78 g/cm3 at 20 °C

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O[C@]1([C@@]2([C@H]([C@H]3[C@@H]([C@@]4(C(=CC(=O)C=C4)CC3)C)C(=O)C2)CC1)C)C(=O)CO

InChI

1S/C21H26O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-15,18,22,26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,18+,19-,20-,21-/m0/s1

InChI key

XOFYZVNMUHMLCC-ZPOLXVRWSA-N

Gene Information

human ... NR3C1(2908)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
PHR1042P29000001559006
format

single component solution

format

neat

format

neat

format

neat

form

liquid

form

-

form

-

form

-

feature

Snap-N-Spike®/Snap-N-Shoot®

feature

-

feature

-

feature

-

packaging

ampule of 1 mL

packaging

-

packaging

-

packaging

-

manufacturer/tradename

Cerilliant®

manufacturer/tradename

-

manufacturer/tradename

EDQM

manufacturer/tradename

USP

General description

Calibration standard and quality control for calibration or identification of prednisone in biological samples for mass spectrometry-based clinical testing applications including therapeutic drug monitoring, endocrinology, cancer treatment, and confirming the cause of secondary adrenal insufficiency. Prednisone is a synthetic corticosteroid drug used to treat a variety of inflammatory diseases and conditions.

Application


  • A Breath of Relief: Oxymetazoline and Flunisolide Nasal Spray in the Management of Myasthenia Gravis Ptosis.: This study explores the use of corticosteroid solutions, like Prednisone, in managing inflammatory conditions. Prednisone solution is essential for understanding its anti-inflammatory effects in clinical settings and can be applied in biochemistry and cell signaling studies (Aloyan et al., 2024).

  • Remission and low disease activity are associated with lower healthcare costs: results from the Systemic Lupus International Collaborating Clinics (SLICC) inception cohort.: Prednisone solution is highlighted for its role in achieving remission and low disease activity in autoimmune diseases like lupus, emphasizing its importance in laboratory and clinical research for autoimmune and inflammatory conditions (Barber et al., 2024).

  • Freeze-dried amniotic membrane graft with a spongy layer in bilateral peripheral ulcerative keratitis: a case report.: This case report discusses the application of Prednisone solution in treating severe inflammatory eye conditions, demonstrating its use in autoimmune disease and inflammatory response research (Bertret et al., 2023).

  • 2023 Canadian Thoracic Society Guideline on Pharmacotherapy in Patients With Stable COPD.: Prednisone solution is part of the pharmacotherapy guidelines for managing chronic inflammatory respiratory conditions like COPD, showcasing its relevance in asthma treatment and biochemistry studies focused on respiratory inflammation (Bourbeau et al., 2023).

  • Retrospective Analysis With Propensity Score Matching of Peripheral T-Cell Lymphoma Treated Frontline With Brentuximab Vedotin and Chemotherapy.: This research highlights the role of Prednisone solution in oncology, specifically in managing inflammatory responses during cancer treatment, making it crucial for pharmacological experimentation in cancer and inflammation research (Burke et al., 2023).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service