Skip to Content
Merck
All Photos(1)

Key Documents

M-141

Supelco

Methylone-D3 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11D3H10NO3 · HCl
CAS Number:
Molecular Weight:
246.71
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); psicótropo (Spain)

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.N(C(C)C(=O)c1cc2c(cc1)OCO2)C([2H])([2H])[2H]

InChI

1S/C11H13NO3.ClH/c1-7(12-2)11(13)8-3-4-9-10(5-8)15-6-14-9;/h3-5,7,12H,6H2,1-2H3;1H/i2D3;

InChI key

GASYWEXOTOXXLK-MUTAZJQDSA-N

General description

A new internal standard suitable for numerous GCMS and LCMS applications from clinical toxicology testing and forensic analysis to urine drug testing and isotope dilution methods. Also known as bk-MDMA or 3,4-methylenedioxy-N-methylcathinone, methylone is a synthetic stimulant of the amphetamine and cathinone classes with significant abuse reported in the UK, Netherlands, Sweden, and New Zealand.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service