Skip to Content
Merck
All Photos(2)

Key Documents

850360P

Avanti

17:0 PC

1,2-diheptadecanoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

PC(17:0/17:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H84NO8P
CAS Number:
Molecular Weight:
762.09
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 200 mg (850360P-200mg)
pkg of 1 × 25 mg (850360P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850360P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCC)=O)=O

InChI

1S/C42H84NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-41(44)48-38-40(39-50-52(46,47)49-37-36-43(3,4)5)51-42(45)35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h40H,6-39H2,1-5H3

InChI key

RTWAYAIMWLNAJW-UHFFFAOYSA-N

General description

Phosphatidylcholines (PCs) are phospholipids, that are abundantly present in the membrane. The fatty acids in the sn-1 and sn-2 positions of PCs are attached to a glycerol molecule covalently with the help of ester bonds.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

17:0 PC (1,2-diheptadecanoyl-sn-glycero-3-phosphocholine) has been used:
  • as an internal standard for phospholipids in fatty acid analysis
  • as a component in the extraction mixture for lipid extraction from frozen leaf tissue
  • as an internal standard and a component in methanol: methyl tert-butyl ether (MTBE) extraction buffer for the determination of metabolite levels in tumor and normal tissues using gas chromatography-mass spectrometrometer (GC-MS)

Biochem/physiol Actions

Phosphatidylcholines (PCs) serve as a major source for eicosanoids.

Packaging

5 mL Amber Glass Screw Cap Vial (850360P-200mg)
5 mL Amber Glass Screw Cap Vial (850360P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piotr Popławski et al.
Biochimica et biophysica acta. Molecular basis of disease, 1863(3), 744-752 (2016-12-26)
Cellular metabolism of renal cell carcinoma (RCC) tumors is disturbed. The clinical significance of these alterations is weakly understood. We aimed to find if changes in metabolic pathways contribute to survival of RCC patients. 35 RCC tumors and matched controls
Kristin A Marks et al.
Genes & nutrition, 8(3), 317-327 (2012-11-28)
Monounsaturated fatty acids (MUFA) have been viewed as either beneficial or neutral with respect to health; however, recent evidence suggests that MUFA may be associated with obesity and cardiovascular disease. Sex differences in MUFA composition have been reported in both
Anthony Sehl et al.
Food & function, 11(2), 1721-1728 (2020-02-11)
The aim of this work was to study the bioavailability of n-3 long-chain polyunsaturated fatty acids (n-3 LC-PUFA), i.e. eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA), carried by marine phospholipids (PL) and formulated in different supramolecular forms. Marine PL were
Dawid Perlikowski et al.
Frontiers in plant science, 7, 1027-1027 (2016-08-04)
Drought tolerant plant genotypes are able to maintain stability and integrity of cellular membranes in unfavorable conditions, and to regenerate damaged membranes after stress cessation. The profiling of cellular glycerolipids during drought stress performed on model species such as Arabidopsis
Kristin A Marks et al.
Prostaglandins, leukotrienes, and essential fatty acids, 89(2-3), 89-96 (2013-06-20)
Sex differences in monounsaturated fatty acid (MUFA) levels suggest ovarian hormones may affect MUFA biosynthesis. Sprague-Dawley rats (8 weeks of age) were ovariectomized or sham operated with ovariectomized rats implanted with a constant-release hormone pellet providing 17β-estradiol, progesterone, both or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service