610007
Avanti
PC Membranes 0.4um
Avanti Research™ - A Croda Brand
Synonym(s):
Extruder PC Membrane, Polycarbonate Membranes 0.4μm 19mm (100/pk)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
40161507
NACRES:
NB.24
manufacturer/tradename
Avanti Research™ - A Croda Brand
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
General description
Polycarbonate Membrane for Avanti′s Mini-Extruder.
Watch the video to learn more about the Avanti Research™ Mini-Extruder.
Watch the video to learn more about the Avanti Research™ Mini-Extruder.
Application
PC Membranes 0.4um has been used to extrude hydrated lipids, for liposome preparation.
Biochem/physiol Actions
Polycarbonate membranes are useful to prepare uniform and specific sized liposomes by ordered extrusion.
Components
Whatman® Nuclepore polycarbonate hydrophilic membrane 10417104
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ionization Properties of Phospholipids Determined by Zeta Potential Measurements
Sathappa M and Alder NN
Bio-protocol, 6(22) (2016)
Preparation of liposomes of defined size distribution by extrusion through polycarbonate membranes
Olson F, et al.
Biochimica et Biophysica Acta - Biomembranes, 557(1), 9-23 (1979)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service