Z743912
ART® SoftFit L Reload System empty reload HLT insert
for 20 ul - 300 ul racks
Synonym(s):
Reload rack, tip rack
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
feature:
for 20 ul - 300 ul racks
packaging:
pkg of 5X10 ea
manufacturer/tradename:
Molecular BioProducts 3704-ER
Recommended Products
feature
for 20 ul - 300 ul racks
packaging
pkg of 5X10 ea
manufacturer/tradename
Molecular BioProducts 3704-ER
technique(s)
PCR: suitable
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The ART SoftFit-L empty racks can be combined with SoftFit-L reload towers and inserts, providing a packaging option that is environmentally friendly and generates less plastic waste compared to standard racks. These racks are designed to be used with ART SoftFit-L reload inserts and reload towers.
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service