Key Documents
Z677493
Camapren chloroprene flock lined gloves
size M
Synonym(s):
reusable glove
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
material
red (chloroprene, natural latex)
red gloves
size
M
sterility
non-sterile
feature
CE compliant
disposable
latex free: no
powder-free
reusable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
standard type EN 421
packaging
package of 100 ea
manufacturer/tradename
KCL 722 - 08
KCL 722
L × thickness
300 mm × 0.6 mm
General description
Camapren flock lined gloves are manufactured from red polychloroprene, a proven material with good, well-balanced properties in relation to chemical and mechanical resistance. A more durable alternative to natural latex. The anatomical design reduces operator fatigue. Good flexibility and a pronounced surface profile ensure excellent sensitivity and a sure grip.
The cotton flock lining improves comfort. Altogether, this is a product range for excellent comfort and wearing characteristics, reliable, economical and popular with users.
  • Ergonomic fit and high elasticity ensures good sensitivity
  • Grooved palm for excellent grip when wet
  • High mechanical resistance
  • Cotton flock lining improves comfort
  • Free from allergenic substances such as thiurames, benzothiazoles and thiourea
  • Protection from radioactive contamination
  • CE compliant
Legal Information
Camapren is a trademark of KCL GmbH