Key Documents
Z676098
IKA® MS 3 minishaker microtiter plate attachment
MS 3.4 microtiter attachment, 1/cs
About This Item
UNSPSC Code:
41103811
NACRES:
NB.73
sterility
non-sterile
packaging
pack of 1
manufacturer/tradename
IKA 0003426400
color
none
General description
Microtiter attachment for holding 1 microtiter plate for use with IKA Twister and MS 3 stirrers.
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)