Key Documents
Z544450
Molymod® molecular model set
teacher/researcher set, organic
About This Item
UNSPSC Code:
60104506
NACRES:
NB.17
feature
no. of atoms 111
no. of bonds 140
manufacturer/tradename
Molymod MMS-003
General description
(38) Black carbon atom parts: (24) tetrahedral, (6) trigonal bipyramidal, (6) trigonal, (2) two-hole linear
(12) Red two-hole angular oxygen atom parts
(40) White single-hole hydrogen atom parts
(4) Blue tetrahedral nitrogen atom parts
(2) Yellow sulfur atom parts: (1) tetrahedral, (1) octahedral
(4) Purple tetrahedral phosphorus atom parts
(8) Green single-hole halogen atom parts
(3) Grey metal atom parts: (2) single-hole, (1) two-hole angular
(18) Lone pair 2D paddles: (6) pink, (6) purple, (6) beige
(55) Medium-length 19mm grey links
(25) Flexible long 30mm grey links
(60) Short 2mm white links
Link remover tool
Storage box
Instructions
Top quality Molymod molecular model sets are for use by students, teachers, and researchers. May be upgraded or customized with additional model parts, for more info, go to sigma-aldrich.com and enter Molymod in our search engine.
Scale is 2.8 cm =100 pm (open style) and 1.45 cm = 100 pm (compact, semi-space-filling). Sets include standard color-coded plastic atom centers (with bond holes set at the appropriate angles), bonding links, and a plastic storage box.
Set pictured is representative of the Student set.
See our Selection Guide for detailed descriptions of set contents.
Legal Information
Molymod is a registered trademark of Spiring Enterprises Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)