Key Documents
Z404942
IKA® IKATRON PT 1000 stainless steel thermometer probes
standard temperature probe, 260 x 3 mm, 1/cs
Synonym(s):
contact thermometer, temperature probe
About This Item
UNSPSC Code:
41112303
NACRES:
NB.76
Pricing and availability is not currently available.
material
stainless steel
packaging
case of 1
pack of 1
manufacturer/tradename
IKA 2735451
L × diam.
260 mm × 3 mm , Depth of immersion: 230 mm
General description
Replacement stainles steel sensor for use with IKA stirrers ETS-D5 and ETS-D6.
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
IKATRON is a trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)