Key Documents
Z281352
Laboratory brushes
Centrifuge, L 12 in., brush diam. × L 1 1/2 in. × 4 1/2 in.
About This Item
UNSPSC Code:
47131605
NACRES:
NB.35
Pricing and availability is not currently available.
packaging
pkg of 4 ea
L
12 in.
brush diam. × L
1 1/2 in. × 4 1/2 in.
Related Categories
General description
Brush materials resistant to acids, aromatic and aliphatic hydrocarbons, ketones, ethyl acetate esters, trichloroethylene, and hot water. FDA and USDA approved materials.
Choose from one of the most recent versions:
Certificates of Analysis (COA)