Key Documents
Z278262
Round bottom flask
capacity 25 mL
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass
round bottom flask
single-neck
sterility
non-sterile
feature
closure type Screw cap
packaging
pack of 1
manufacturer/tradename
Ace Glass 9592-15
capacity
25 mL
General description
With cap, FETFE® O-ring, and septum. Threaded <ST/>14/10 joint.
Note: This product is part of a kit. Please refer to the Z510297 web page for additional information.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)