Key Documents
Z222380
Metal standard taper clip
joint size, 24
Synonym(s):
Metal clip, Taper clip
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
General description
Nickel-plated spring-steel joint clips offer increased strength and resistance to heat and cracking over Delrin® plastic type. Securely holds conical joints under pressure at temperatures up to 392 °F (200 °C). Will not scratch glassware.
Legal Information
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)