Key Documents
Z219290
Sigma-Aldrich
Plug wrench
bronze alloy, pkg of 1 ea
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
Pricing and availability is not currently available.
packaging
pkg of 1 ea
General description
Non-sparking bronze alloy. Fits the most popular bung patterns.
Choose from one of the most recent versions:
Certificates of Analysis (COA)