Key Documents
Z102466
Luer-to-Luer connector
male Luer lock to male Luer lock, nickel plated
Synonym(s):
Luer connector, Luer fitting
About This Item
UNSPSC Code:
41122000
NACRES:
NB.03
Pricing and availability is not currently available.
material
nickel plated
manufacturer/tradename
Popper 6177
Related Categories
General description
Water tested 45 psi.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Can this product be autoclaved (121oc)?
    1 answer
    1. This material can be autoclaved.
      (This material is not the following: As a general rule of thumb, you CANNOT autoclave materials that are contaminated with solvents, radioactive materials, volatile or corrosive chemicals, or items that contain mutagens, carcinogens, or teratogens)
      Helpful?